missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Applied Biosystems™ RapidFinder™ STEC Screening Assay
RapidFinder™ STEC Screening Assay
Brand: Applied Biosystems™ 4476886
10818.00 DKK valid until 2025-08-24
Use promo code "24117" to get your promotional price.
This item is not returnable.
View return policy
Description
The Applied Biosystems™ RapidFinder™ STEC Screening Assay is part of a complete, validated screening and confirmation workflow enabling TaqMan™ real-time PCR detection of the Shiga toxin-producing E. coli (STEC) O157:H7 and Big 6 non‐O157:H7 serogroup (O26, O45, O103, O111, O121 and O145) in a variety of beef products. The RapidFinder STEC Screening Assay was co-developed with the USDA-ARS and contains 96 complete qPCR assays targeted to screen for Shiga toxin 1 and 2 (stx1/stx2), eae, and potential O157:H7 presence. The assay is designed to be inclusive of all know allelic variants of stx and eae, including stx2g and stx2f. All reactions include an internal positive control (IPC) and ROX™ passive reference. Samples assessed as negative require no further testing. Samples assessed as positive must be further tested using the RapidFinder™ STEC Confirmation Assay.The RapidFinder STEC Detection Workflow provides:
Studies have shown that the RapidFinder STEC assays have a high level of sensitivity and specificity for E. coli O157:H7 and the Big 6 non-O157 STEC strains*. They detected all inclusion strains and showed no cross-reactivity to any of the exclusion strains tested. The stx assays detected all known variants of stx1 and stx2, including stx2f and stx2g. The optimized workflow co-developed with USDA-ARS showed equivalent detection to the MLG reference method, and the internal positive control in both assays significantly reduces the risk of false-negatives.
Applications
The RapidFinder STEC Detection Workflow is intended for use by microbiological analysts who need to test for STEC in beef samples. Combined with the Applied Biosystems 7500 Fast RT-PCR System, RapidFinder STEC screening and confirmation assays provide a comprehensive solution for rapid detection, identification, and confirmation of Big 6 STECs and E. coli 0157:H7 to meet importation and exportation requirements for the beef industry. Specific workflows are detailed in the figures below and include both automated and spin column-based DNA isolation and real-time PCR detection of E. coli O157:H7 and Big 6 non-O157 strains in beef samples.
RapidFinder Express Software
When integrated into the RapidFinder STEC Detection Workflow, RapidFinder Express Software automates data interpretation and generates user instructions on how to proceed with a sample. Data displays are available for novice and advanced users, showing simple presence/absence calls or raw data files using 7500 Fast System SDS Software.
RapidFinder STEC Screening and Confirmation Assays meet USDA FSIS guidance
The RapidFinder STEC Detection Workflow, a two-stage real-time PCR method, is designed to accurately detect, identify, and confirm E. coli O157:H7 and the Big 6 serogroups of non-O157 Shiga toxin-producing E. coli defined by the USDA Food Safety and Inspection Service (FSIS) as adulterants in the American beef industry.
Learn more about the RapidFinder STEC Detection Workflow >
*Swimley, Michelle S. Design and Evaluation of Two-Stage Multiplex Real-Time PCR Method for Detecting O157:H7 and non-O157 STEC Strains from Beef Samples. IAFP 2015.

Specifications
One 96-well plate loaded with 12 x 8-tube strips containing a total of 96 lyophilized qPCR reactions and one negative control box. 2 to 8°C | |
Primer-probe | |
96 rxn(s) | |
96 Reactions |
RapidFinder™ STEC Screening Assay, 96 rxns, Primer-Probe Detection Method, Store at 2 to 8°C, Wet ice Shipping, For Food and Beverage, Meat Testing | |
RapidFinder™ STEC Screening Assay | |
Food and Beverage | |
96 rxn(s) |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
For Testing of Food and Environmental Samples Only.
Spot an opportunity for improvement?Share a Content Correction