missing translation for 'onlineSavingsMsg'
Learn More
Learn More
RAB43 Mouse anti-Human, Clone: 3B2A8, Proteintech
Mouse Monoclonal Antibody
Marca: Proteintech 66654-1-IG-20UL
Questo articolo non è restituibile.
Consulta la politica di reso
Descrizione
The small GTPases Rab are key regulators of intracellular membrane trafficking, from the formation of transport vesicles to their fusion with membranes. Rabs cycle between an inactive GDP-bound form and an active GTP-bound form that is able to recruit to membranes different set of downstream effectors directly responsible for vesicle formation, movement, tethering and fusion. The low intrinsic GTPase activity of RAB43 is activated by USP6NL. Involved in retrograde transport from the endocytic pathway to the Golgi apparatus. Involved in the transport of Shiga toxin from early and recycling endosomes to the trans-Golgi network. Required for the structural integrity of the Golgi complex. Plays a role in the maturation of phagosomes that engulf pathogens, such as S.aureus and M.tuberculosis.Specifica
RAB43 | |
Monoclonal | |
1 mg/mL | |
PBS with 50% glycerol and 0.1% sodium azide; pH 7.3 | |
Q86YS6 | |
RAB43 | |
RAB43 Fusion Protein Ag26911 | |
20 μL | |
Primary | |
Human | |
Antibody | |
IgG1 |
Western Blot | |
3B2A8 | |
Unconjugated | |
RAB43 | |
ISY1, RAB11B, RAB41, RAB43, Ras related protein Rab 41, Ras related protein Rab 43 | |
Mouse | |
Protein G | |
RUO | |
339122 | |
-20°C | |
Liquid |
Correzione del contenuto del prodotto
Fornite il vostro feedback sul contenuto del prodotto compilando il modulo sottostante.
Titolo del prodotto
Individuate un'opportunità di miglioramento?Condividi una correzione di contenuto