Learn More
Quickfit™ Glass Conical Funnel with Semi-Micro Glass Sintered Disc
Marca: Scilabware SF4T30
Questo articolo non è restituibile.
Consulta la politica di reso
Descrizione
- Ideal for filtering semi-micro quantities of material
- Fitted with borosilicate glass sintered disc
- Suitable for use with Quickfit™ MD range of receivers
Specifica
Borosilicate Glass | |
Quickfit™ MD: range of receivers | |
Sintered glass disc | |
Semi-micro filter funnel |
10/19 | |
10mm | |
3 |