Learn More
Karl Hecht™ PTFE Sleeves for Ground Joints and Stoppers
A disposable option for preventing ground joints from sticking together.
Marca: Karl Hecht™ 42388019
Questo articolo non è restituibile.
Consulta la politica di reso
Descrizione
- Self-lubricating
- Thickness of approx. 0.05mm
- For use with joint size 19/26
- Outstanding chemical resistance at temperatures from −200° to +260°C
- Provided in packs of 10
Specifica
Prevents ground joints from sticking | |
PTFE | |
−200° to +260°C | |
19/26 |
Joint size 19/26 | |
10/pk | |
Approx. 0.05 mm | |
Protecting Sleeve |