Learn More
PTBP1 Mouse anti-Human, Unconjugated, Clone: 3D6D8, Proteintech
Mouse Monoclonal Antibody
Marca: Proteintech 67462-1-IG-150UL
Questo articolo non è restituibile.
Consulta la politica di reso
Descrizione
The PTBP1 antibody from Proteintech is a mouse monoclonal antibody generated with recombinant protein of human PTBP1. This antibody recognizes human, mouse, rat antigen. The PTBP1 antibody has been validated for the following applications: IF, IHC, WB, ELISA analysis.
Specifica
PTBP1 | |
Monoclonal | |
1000 μg/mL | |
Mouse | |
Primary | |
Human, Rat, Mouse |
Western Blot, Immunohistochemistry, Immunofluorescence, ELISA | |
3D6D8 | |
Unconjugated | |
150 μL | |
5725 | |
IgG1 |