Learn More
Kimberly-Clark™ Kimtech™ Procedure Masks
Non-sterile pleat-style masks with high bacterial and particle filtration, and certified to EN14683 Type II. The latex- and silicone-free masks reduce contamination while ensuring comfort, with a low-lint outer layer and adjustable nose seal.
Specifica
Size | Standard |
---|---|
Sterilità | Non-sterile |
Stile | Pleated |
Certificazione/Conformità | ISO 14001, ISO 9001; Industrial Use Only |
Senza lattice | Yes |
Descrizione
- Pleat style masks with earloops
- Excellent particle and bacterial filtration efficiency to ensure the wearer’s security.
- Certified according to EN14683 Type II, and provided with supporting technical information to demonstrate regulatory compliance.
- Constructed from a low-lint polyethylene film outer layer for enhanced protection.
- Contain no natural rubber latex or silicone, reducing the risks of skin irritation for the wearer.
- The masks are provided in a single universal size.
Specifica
Standard | |
Pleated | |
Yes |
Non-sterile | |
ISO 14001, ISO 9001; Industrial Use Only |