Learn More
Crison™ GLP 22 pH and Ion Meter
Combining an excellent pH meter and a large ionometer, the GLP 22 is ideal for research laboratories.
Brand:  Crison™ 2200
Product Code. 12001282
  • 2240.00€ / Each
Estimated Shipment: 12-06-2025
Explore more special offers
Add to basket
Description
Description
  • 2 measuring channels for pH, redox, or ISE
  • Able to manage 2 magnetic stirrers
  • Data logger where the last 400 measurements per channel are stored, calibration data and electrode history
  • pH resolution selectable between 0.1, 0.01 and 0.001 pH
  • Several calibration modes: with recognition of 3 families of buffers, at a specific pH, with user buffers, etc.
  • 3 measurement modes: stability, continuous, or time
  • As an ionometer, up to 4 calibration curves per channel, calibration with 2 to 5 patterns programmable by user, and selectable calibration units mol/L, g/L, mmol/L or mg/L
  • Can be connected to a PC, printer, external PC keyboard or barcode reader
  • Magnetic agitator controlled automatically by the instrument
  • Support for the electrode with location to hold the cable and prevent breakage
  • Large graphic screen with clear messages that guide the user at all times
  • Keyboard of great industrial quality and maximum simplicity
  • Bottles for calibration with stirring magnet, more than 10 calibrations with only 40mL of buffer
Specifications
Specifications
<0.005
-2 to 16
0.001, 0.01, 0.1
±2000
Product Suggestions
Product Suggestions
Videos
SDS
Documents
Documents
Certificates
Special Offers
Special Offers