Learn More
Fisherbrand™ PE Plug for Shell Vial
Polyethylene caps are chemically inert and suitable for most chromatography applications.
3.35 EUR - 32.84 EUR
Specifications
Color Transparent
Material Polyethylene
Quantity 100
Type Plug
View More Specs
Products 4
Product Code Brand Diameter (Metric) Outer dimClosureSize Price Quantity & Availability  
Product Code Brand Diameter (Metric) Outer dimClosureSize Price Quantity & Availability  
11804960
View Documents Promotion Details
Fisherbrand™
12 08 0546
12 mm
32.35 EUR
Pack of 1000
Only null left
Estimated Shipment: 14-07-2025
Add to basket
11556014
View Documents
Fisherbrand™
12 08 0546
12 mm This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
N/A
11997061
View Documents
Fisherbrand™
08 08 1167
8 mm This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
N/A
11824980
View Documents
Fisherbrand™
15 08 0547
15 mm This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
N/A
Description
Description
  • PE Plug
  • Transparent
  • Snap caps are easy to apply and easy to remove, eliminate the need for crimping or de-capping tools
Specifications
Specifications
Transparent
Polyethylene
100
Plug