Learn More
Ansell™ NitraSafe™ 28-360 Series Nitrile Heavy Weight Gloves
Sfoglia Tutto Ansell Prodotti
Click to view available options
Size:
8
9
10
Descrizione
- Industrial gloves
- NitraSafe™ range stands for safe handling of oily heavy metal parts
- Mechanical protection against cuts and abrasion
- Heavy weight design provides feeling of cushioning for the wearer
- Protective gloves combines the protection of Kevlar™ reinforced construction and grip, flexibility of advanced nitrile coating technology
Applications:
- Handling parts with sharp, rough edges
Specifica
Specifica
Materiale parte posteriore | Nitrile Foam |
Certificazione/Conformità | CE Mark 0493, Standard Category III EN 388 : 3321 |
Materiale polso | Nitrile |
Product Type | Heavyweight Gloves |
Size | 8 |
Stile polso guanto | Nitragold™ |
Materiale rivestimento interno | Cotton Jersey Dupont™ Kevlar™ aramid fiber |
Rivestimento interno | Cotton Jersey Dupont™ Kevlar™ aramid fiber |
Tipo di palmo | Fully coated-palm |
Quantità | 72 Pairs |