Learn More
Nickel Electro™ Nickel Crucible Lid
Complete a Nickel Crucible with a flat lid that has a depression for centering and an extension on one side that serves as a handle. Nickel Electro™ Nickel Crucible Lid comes in a range of sizes to fit the dimensions of the crucible being used.
Marca: Nickel Electro™ 46
Includes: DiameterMetricOuter
Requires : Autoclavable,dimMaterial,ProductTypeSpecs
Questo articolo non è restituibile. Consulta la politica di reso
Descrizione
- Resistant to corrosion
- Should not be used with nitric acid
- Prevents chance of hot material spattering out of open crucible
- Cost effective replacement for platinum
Crucible lid, nickel, handle
Specifica
Nickel | |
Autoclavable | |
Crucible lid |
Each | |
30 mm |