Learn More
Thermo Scientific™ Kit preassemblato Target DP™ National
Thermo Scientific™ National Preassembled Target DP Kits include 100 vials with pre-attached caps and septa.
Marca: Thermo Scientific™ C4000-188WR
Questo articolo non è restituibile.
Consulta la politica di reso
Descrizione
Thermo Scientific™ National Preassembled Target DP Kits include 100 vials with pre-attached caps and septa.
- Packaged in convenient vial trays with clear covers or in polybags
Specifica
Red | |
Amber Glass I-D; Red | |
Glass | |
PTFE/Silicone/PTFE | |
Sì |
Red/White/Red | |
Amber | |
2 mL | |
9-425 |