missing translation for 'onlineSavingsMsg'
Learn More
Learn More
MYOT Rabbit anti-Human, Mouse, Rat, Polyclonal, Proteintech
Rabbit Polyclonal Antibody
Marca: Proteintech 10731-1-AP-20UL
Questo articolo non è restituibile.
Consulta la politica di reso
Descrizione
This antibody can detect both of isoforms around 57 kDa and 35 kDa.
This gene encodes a cystoskeletal protein which plays a significant role in the stability of thin filaments during muscle contraction. This protein binds F-actin, crosslinks actin filaments, and prevents latrunculin A-induced filament disassembly. Mutations in this gene have been associated with limb-girdle muscular dystrophy and myofibrillar myopathies. Several alternatively spliced transcript variants of this gene have been described, but the full-length nature of some of these variants has not been determined.Specifica
MYOT | |
Polyclonal | |
Unconjugated | |
MYOT | |
57 kDa cytoskeletal protein, LGMD1, LGMD1A, MYOT, myotilin, TTID | |
Rabbit | |
Antigen Affinity Chromatography | |
RUO | |
291605, 58916, 9499 | |
-20°C | |
Liquid |
Immunohistochemistry (Paraffin), Western Blot | |
0.18 mg/mL | |
PBS with 50% glycerol and 0.1% sodium azide; pH 7.3 | |
Q9JIF9, Q9UBF9 | |
Myot | |
MYOT Fusion Protein Ag1112 | |
20 μL | |
Primary | |
Mouse, Rat, Human | |
Antibody | |
IgG |
Correzione del contenuto del prodotto
Fornite il vostro feedback sul contenuto del prodotto compilando il modulo sottostante.
Titolo del prodotto
Individuate un'opportunità di miglioramento?Condividi una correzione di contenuto