missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ Monkey PF-4 ELISA Kit
ELISA
Specifica
Numero di accesso | chemokine (C-X-C motif) ligand 4; C-X-C motif chemokine 4; CXCL4; Endothelial cell growth inhibitor; Iroplact; Oncostatin-A; Pf4; PF-4; Pf4a; Platelet factor 4; Platelet factor 4, short form; RATPF4A; Scyb4 |
---|---|
Intervallo saggio | 0.041 to 10 ng/mL |
Assay Sensitivity | 0.041 ng/mL |
Coniugato | HRP |
Product Type | ELISA |
Descrizione
Monkey PF-4 quantitates monkey PF-4 in serum, plasma, supernatant. The assay will exclusively recognize both natural and recombinant monkey PF-4.
Platelet factor-4 is a 70-amino acid protein that is released from the alpha-granules of activated platelets and binds with high affinity to heparin. Its major physiologic role appears to be neutralization of heparin-like molecules on the endothelial surface of blood vessels, thereby inhibiting local antithrombin III activity and promoting coagulation. As a strong chemoattractant for neutrophils and fibroblasts, PF4 probably has a role in inflammation and wound repair (Eisman et al., 1990).Specifica
chemokine (C-X-C motif) ligand 4; C-X-C motif chemokine 4; CXCL4; Endothelial cell growth inhibitor; Iroplact; Oncostatin-A; Pf4; PF-4; Pf4a; Platelet factor 4; Platelet factor 4, short form; RATPF4A; Scyb4 | |
0.041 ng/mL | |
ELISA | |
Colorimetric Microplate Reader | |
<10 | |
HRP | |
Plasma 50 μL, Serum 50 μL, Supernatant 50 μL | |
PF4 | |
4 hrs 45 mins |
0.041 to 10 ng/mL | |
HRP | |
Plasma, Serum, Supernatant | |
<12% | |
Pre-coated 96 well plate, Standard, Assay Diluent concentrate, Biotinylated Detection Antibody, SAV-HRP, Wash Buffer, Chromogen, Stop Solution, Adhesive Plate Covers | |
RUO | |
2°C to 8°C | |
1 hrs 20 mins |
Individuate un'opportunità di miglioramento?Condividi una correzione di contenuto
Correzione del contenuto del prodotto
Fornite il vostro feedback sul contenuto del prodotto compilando il modulo sottostante.
Titolo del prodotto