missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ Monkey IL-13 ELISA Kit
Monkey IL-13 quantitates monkey IL-13 in serum, plasma, supernatant.
566.00€ - 2105.00€
Specifications
Accession Number | ALRH; ALRH antibody; BHR1; il 13; Il13; Il-13; IL-13; cytokine; ILN; Interleukin; interleukin 13; Interleukin13; Interleukin-13; interleukin-13 precursor; L13; MGC116786; MGC116788; MGC116789; NC30; NC30 antibody; P600; P600 antibody; T-cell activation protein P600 |
---|---|
Assay Range | 0.05 to 10 ng/mL |
Assay Sensitivity | 0.05 ng/mL |
Conjugate | Biotin |
Product Type | ELISA |
Description
Monkey IL-13 quantitates monkey IL-13 in serum, plasma, supernatant. The assay will exclusively recognize both natural and recombinant monkey IL-13.
IL-13 is an immunoregulatory cytokine produced primarily by activated Th2 cells, and is involved in regulating inflammatory and immune responses. IL-13 down-regulates macrophage activity, thereby inhibiting the production of pro-inflammatory cytokines and chemokines, and is involved in allergen-induced asthma through mechanisms independent of IgE and eosinophils. IL-13 also regulates several stages of B cell maturation and differentiation, up-regulates CD23 and MHC class II expression, and promotes IgE isotype switching of B cells. Structurally, IL-13 closely resembles IL-3, IL-4, IL-5 and GM-CSF. The gene encoding IL-13, IL3, IL5, IL4, and CSF2 form a cytokine gene cluster on chromosome 5q, with the IL-13 gene particularly close to IL4.Specifications
ALRH; ALRH antibody; BHR1; il 13; Il13; Il-13; IL-13; cytokine; ILN; Interleukin; interleukin 13; Interleukin13; Interleukin-13; interleukin-13 precursor; L13; MGC116786; MGC116788; MGC116789; NC30; NC30 antibody; P600; P600 antibody; T-cell activation protein P600 | |
0.05 ng/mL | |
ELISA | |
Colorimetric Microplate Reader | |
<10% | |
HRP | |
Supernatant, 50 μL; Plasma, 50 μL; Serum, 50 μL | |
IL-13 | |
4 hr 45 min |
0.05 to 10 ng/mL | |
Biotin | |
Plasma, Serum, Supernatant | |
<12% | |
Pre-coated 96 well plate, Standard, Assay Diluent concentrate, Biotinylated Detection Antibody, SAV-HRP, Wash Buffer, Chromogen, Stop Solution, Adhesive Plate Covers | |
Research Use Only | |
2°C to 8°C | |
1 hr 20 min |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title