Learn More
Jenway™ Model 4510 and Model 4520 Benchtop Conductivity Meter
Designed to provide accurate and quick readings.
Marca: Jenway™ 35440-10
Questo articolo non è restituibile.
Consulta la politica di reso
Descrizione
- Always measure at the best resolution with automatic standard recognition
- Easily calibrate with either standard solution or known cell constant
Specifica
1413 μS, 12.88 mS, 84 μS, 10 μS, 1382 ppm TDS | |
Conductivity, TDS, Resistivity, Salinity, Temperature | |
Glass conductivity probe with ATC (K = 1) as noted, electrode holder, and GLP | |
Conductivity Probe |
K = 1 | |
0 to 19.99 μS / 0 to 199.9 μS / 0 to 1999 μS / 0 to 19.99 mS / 0 to 199.9 mS / 0 to 1999 mS / 0 to 19.99 S*(only with cell constant >5) | |
English, French, German, Italian, Spanish, Portuguese | |
0.01 μS / 0.1 μS / 1 μS / 0.01 mS / 0.1 mS / 1 mS / 0.01 S |
Titolo del prodotto
Jenway™ Model 4510 and Model 4520 Benchtop Conductivity Meter > Includes: Glass conductivity probe with ATC (K = 1) as noted, electrode holder, and GLP