Learn More
3M™ Maxim™ RAS Safety Glasses
Click to view available options
:
Lens Tint: Trasparente
Descrizione
- Safety spectacles with a super hard eyewear coating that provides excellent protection against scratches in the toughest of environments
- RAS coating 5 times more scratch resistance than generic hard coats
- RAS coating fulfils the EN 166 requirements for K marking
- Spectacles with ventilation channel across the lens to minimize misting
- Soft, adjustable nose bridge and a soft-touch brow guard
- Adjustable temple lengths and pantoscopic (angle) adjustments
- Design provides excellent coverage and good field of vision
Specifica
Specifica
Lens Coating | Scratch-resistant |
Lens Tint | Clear |
Product Type | Safety glasses |
Colore telaio | Black |