Learn More
Ansell Edmont™ Marigold™ Blue Natural Rubber Latex Immersion Gloves
Sfoglia Tutto Ansell Prodotti
Click to view available options
Size:
7
8
9
10
11
Descrizione
- Crinkle finish provides grip in wet and dry environments
- Polyamide seamless liner
- Wrinkle natural rubber latex coating
- Gauntlet cuff
Specifica
Specifica
Materiale parte posteriore | Rubber Latex |
Certificazione/Conformità | Standards EN 374, EN 407, EN 388 : 2241, 21 CFR (USA), CFIA (Canada) |
Materiale polso | Natural Rubber Latex |
Coating Material | Natural Rubber Latex |
Latex Free | No |
Product Type | Immersion Gloves |
Size | 11 |
Spessore (sistema imperiale) | 50 mil |
Stile polso guanto | Elasticated |
Rivestimento interno | Natural Rubber Latex |