Learn More
Ansell KleenGuard™ Unispec™ II Safety Glasses
Click to view available options
Lens Tint:
Clear
Unit Size:
Pack of 50
Description
- Low-cost wraparound protection with one-piece clear polycarbonate lens
- Uncoated clear frame
- Side shields provide added protection
Compliance
Meets ANSI Z87.1+ 2010 standards
Specifications
Specifications
Certifications/Compliance | CE Approved, ANSI Z87.1/CSA Z94.3 |
Lens Coating | Uncoated |
Lens Tint | Clear |
Product Type | Safety Glasses |
Material | Plastic |
Disposable | Reusable |
Lens Material | Plastic |
Frame Material | Plastic |
UV Protection | UV400 |
Frame Type | Frameless |