Learn More
KCTD2/5/17 Rabbit anti-Human, Unconjugated, Polyclonal, Proteintech
Rabbit Polyclonal Antibody
Marca: Proteintech 15553-1-AP-150UL
Descrizione
The KCTD2/5/17 antibody from Proteintech is a rabbit polyclonal antibody generated with recombinant protein of human KCTD2/5/17. This antibody recognizes human, mouse, rat antigen. The KCTD2/5/17 antibody has been validated for the following applications: FC, IF, IHC, IP, WB, ELISA analysis.
Specifica
KCTD2/5/17 | |
Polyclonal | |
Unconjugated | |
150 μL | |
54442 | |
IgG |
Western Blot, Immunoprecipitation, Immunohistochemistry, Immunofluorescence, Flow Cytometry | |
333 μg/mL | |
Rabbit | |
Primary | |
Human, Rat, Mouse |