Learn More
Brady™ ISO 7010 Sign - Wear head protection
Prevent costly accidents with ISO 7010 safety signs for optimal impact and recognizability.
Marca: Brady™ 819915
Questo articolo non è restituibile.
Consulta la politica di reso
Descrizione
- Polypropylene
- For indoor and outdoor use
Specifica
ISO 7010 | |
Polypropylene | |
Indoor and Outdoor | |
-40°C to 120°C | |
Global |
PIC M014-Dia 200-PP-CRD/1 | |
1 Ea. | |
1.501 mm | |
Round | |
White on Blue |