Learn More
Iron(III) ethoxide, soluble polymeric mixture, Thermo Scientific™
Codice prodotto. 15482467
Click to view available options
Quantità:
2 g
10 g
50 g
This Thermo Scientific brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.
TRUSTED_SUSTAINABILITY
Identificatori chimici
CAS 5058-42-4
Formula molecolare C6H15FeO3
Molecular Weight (g/mol) 191.028
Numero MDL MFCD00078028
InChI Key JTPUGUWXHGEEHW-UHFFFAOYSA-N
Sinonimo iron iii ethoxide, iron 3+ ethanolate, triethoxyiron iii, ethanolate; iron 3+, acmc-1alnr, iron 3+ tris ethoxide, iron 3+ ion tris ethoxide, iron iii ethoxide trace metals basis 500mg
PubChem CID 6452222
IUPAC Name ethanolate;iron(3+)
SMILES CC[O-].CC[O-].CC[O-].[Fe+3]
Specifica
Materiale o nome chimico Iron (III) ethoxide
Nota nome Soluble polymeric mixture
Colore Brown
Punti di ebollizione (decomposition)
Quantità 10 g
Numero UN UN2926
Informazioni di solubilità Soluble in hexane and ethanol
Forma fisica Solid