Learn More
Smeg™ Injection Trolley for Laboratory Glassware Washers
Use these trolleys with Smeg™ glassware washers. Smeg Injection Trolleys for Laboratory Glassware Washers are available in a variety of configurations to fit a wide range of applications.
Marca: Smeg™ LM24
Questo articolo non è restituibile.
Consulta la politica di reso
Descrizione
- For use with Smeg glassware washers
- Available with one, two and mixed spraying/injection levels
- Clean and sanitize a wide variety of labware
Specifica
Stainless Steel | |
24 items | |
Injection trolley with one injection level | |
Glassware washers | |
1 Each |
Injection Trolley | |
Capacity of 24 items; can wash both large items and small flasks or bottles. Maximum height of glassware is 500mm | |
Washing narrow-necked glassware of different sizes | |
Fittings for ULB 40 adaptor nozzles for pipettes |