Learn More
Fisherbrand™ Individual Stainless Steel Leaf Calibration Weights, Class E2 with certificate
Calibrate weights of lower accuracy classes. Fisherbrand™ Class E2 Individual Stainless Steel Leaf Calibration Weights are intended for use in the verification or calibration of class F1 weights and for use with weighing instruments of special accuracy class I.
Marca: Fisherbrand™ FB67820
60.15 EUR valido fino al 2025-12-31
Usa il codice promo "25339" per avere il tuo prezzo promozionale.
Descrizione
- Class E2 calibration weight
- In compliance with OIML R111
- Delivered with a calibration certificate guaranteeing traceability
Specifica
E2 | |
Stainless Steel | |
500 mg | |
500 mg |