Learn More
Ansell HyFlex™ 48-101 Polyurethane Coated Seamless Knit Nylon Gloves
Gloves are designed for light applications that require precise handling of small objects in dry or slightly oily environments.
Marca: Ansell 48101070
26.62 EUR valido fino al 2025-12-31
Usa il codice promo "25339" per avere il tuo prezzo promozionale.
Descrizione
- Designed for light duty applications that require comfort and abrasion resistance
- Polyurethane coating enhances grip in dry or light oil applications
- Seamless construction prevents irritation
Specifica
ANSI 4 | |
Standard CATEGORY II EN 388 : 4131 | |
Knit | |
Polyurethane | |
Rough | |
Polyurethane, Nylon | |
7 | |
Nero | |
Rough | |
Coated-palm | |
Abrasion resistance |
Polyurethane | |
No | |
Nylon | |
Black | |
Specific | |
Lightweight Gloves | |
Unisex | |
Knit | |
Nylon | |
12 paia | |
48-101 |