missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ Human Syndecan-4 ELISA Kit
ELISA
Specifica
Numero di accesso | AA959608; Amphiglycan; AW108331; MGC22217; RATRYUDOCA; RYUDOCA; ryudocan; ryudocan amphiglycan; ryudocan core protein; Ryudocan/syndecan 4; S4; Sdc4; SYND4; syndecan 4; syndecan 4 (amphiglycan, ryudocan); syndecan proteoglycan 4; syndecan-4 |
---|---|
Intervallo saggio | 6.14 to 1,500 pg/mL |
Assay Sensitivity | 6 pg/mL |
Coniugato | HRP |
Product Type | ELISA |
Descrizione
Human Syndecan-4 quantitates human Syndecan-4 in serum, plasma, supernatant. The assay will exclusively recognize both natural and recombinant human Syndecan-4.
Syndecans are type I integral membrane proteoglycans that are involved in cell-extracellular matrix adhesion and growth factor binding. Syndecan 1 is a matrix receptor which binds to Collagens, Fibronectin and Thrombospondin. Syndecan 1 and Syndecan 3 interact with MK (midkine), a growth/differentiation factor involved in embryogenesis. Syndecan 2 is highly expressed at areas of high morphogenetic activity, such as epithelial-mesenchymal interfaces and the prechondrogenic and preosteogenic mesenchymal condensations. Syndecan 4 functions cooperatively with integrins in the processes of cell spreading and focal adhesion assembly.Specifica
AA959608; Amphiglycan; AW108331; MGC22217; RATRYUDOCA; RYUDOCA; ryudocan; ryudocan amphiglycan; ryudocan core protein; Ryudocan/syndecan 4; S4; Sdc4; SYND4; syndecan 4; syndecan 4 (amphiglycan, ryudocan); syndecan proteoglycan 4; syndecan-4 | |
6 pg/mL | |
ELISA | |
Colorimetric Microplate Reader | |
Sdc4 | |
<10 | |
HRP | |
Plasma 50 μL, Serum 50 μL, Supernatant 50 μL | |
Syndecan 4 | |
4 hrs 45 mins |
6.14 to 1,500 pg/mL | |
HRP | |
Plasma, Serum, Supernatant | |
6385 | |
<12% | |
Pre-coated 96 well plate, Standard, Assay Diluent concentrate, Biotinylated Detection Antibody, SAV-HRP, Wash Buffer, Chromogen, Stop Solution, Adhesive Plate Covers | |
RUO | |
2°C to 8°C | |
1 hrs 20 mins |
Individuate un'opportunità di miglioramento?Condividi una correzione di contenuto
Correzione del contenuto del prodotto
Fornite il vostro feedback sul contenuto del prodotto compilando il modulo sottostante.
Titolo del prodotto