missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ Human Siglec-6 ELISA Kit
ELISA
Specifica
Numero di accesso | CD327; CD33 antigen-like 1; CD33L; CD33L1; CD33L2; CDW327; OBBP1; OB-BP1; Obesity-binding protein 1; sialic acid binding Ig like lectin 6; sialic acid binding Ig-like lectin 6; sialic acid-binding Ig-like lectin 6; Siglec; SIGLEC6; Siglec-6 |
---|---|
Intervallo saggio | 0.05 to 10 ng/mL |
Assay Sensitivity | 0.05 ng/mL |
Coniugato | HRP |
Product Type | ELISA |
Descrizione
Human Siglec-6 quantitates human Siglec-6 in serum, plasma, supernatant. The assay will exclusively recognize both natural and recombinant human Siglec-6.
SIGLEC9 (Sialic acid-binding Ig-like lectin 9) is a putative adhesion molecule that mediates sialic-acid dependent binding to cells. It contains two Ig-like C2 type (immunoglobulin-like) domains and one Ig-like V-type (immunoglobulin-like) domain. It preferentially binds to alpha2, 3-or alpha2, 6-linked sialic acid. The sialic acid recognition site may be mased by cis interactions with sialic acids on the same cell surface.Specifica
CD327; CD33 antigen-like 1; CD33L; CD33L1; CD33L2; CDW327; OBBP1; OB-BP1; Obesity-binding protein 1; sialic acid binding Ig like lectin 6; sialic acid binding Ig-like lectin 6; sialic acid-binding Ig-like lectin 6; Siglec; SIGLEC6; Siglec-6 | |
0.05 ng/mL | |
ELISA | |
Colorimetric Microplate Reader | |
SIGLEC6 | |
<10 | |
HRP | |
Plasma 50 μL, Serum 50 μL, Supernatant 50 μL | |
SIGLEC6 | |
4 hrs 45 mins |
0.05 to 10 ng/mL | |
HRP | |
Plasma, Serum, Supernatant | |
946 | |
<12% | |
Pre-coated 96 well plate, Standard, Assay Diluent concentrate, Biotinylated Detection Antibody, SAV-HRP, Wash Buffer, Chromogen, Stop Solution, Adhesive Plate Covers | |
RUO | |
2°C to 8°C | |
1 hrs 20 mins |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title