missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ Human Notch-3 ELISA Kit
ELISA
Specifica
Numero di accesso | AW229011; CADASIL; CADASIL1; CASIL; hpbk; IMF2; LMNS; N3; neurogenic locus notch homolog protein 3; Notch 3; Notch 3 extracellular truncation; Notch 3 intracellular domain; Notch gene homolog 3; Notch homolog 3; Notch3 |
---|---|
Intervallo saggio | 0.05 to 10 ng/mL |
Assay Sensitivity | 0.05 ng/mL |
Coniugato | HRP |
Product Type | ELISA |
Descrizione
Human Notch-3 quantitates human Notch-3 in serum, plasma, supernatant. The assay will exclusively recognize both natural and recombinant human Notch-3.
NOTCH3 is the third discovered human homologue of the Drosophilia melanogaster type I membrane protein notch. In Drosophilia, notch interaction (with its cell-bound ligands delta, serrate) establishes an intercellular signalling pathway that plays a key role in neural development. Homologues of the notch-ligands have also been identified in human. NOTCH3 functions as a receptor for membrane-bound ligands Jagged1, Jagged2, and Delta1 to regulate cell-fate determination. Mutations in NOTCH3 have been identified as the underlying cause of cerebral autosomal dominant arteriopathy with subcortical infarcts and leukoencephalopathy (CADASIL).Specifica
AW229011; CADASIL; CADASIL1; CASIL; hpbk; IMF2; LMNS; N3; neurogenic locus notch homolog protein 3; Notch 3; Notch 3 extracellular truncation; Notch 3 intracellular domain; Notch gene homolog 3; Notch homolog 3; Notch3 | |
0.05 ng/mL | |
ELISA | |
Colorimetric Microplate Reader | |
Notch3 | |
<10 | |
HRP | |
Plasma 50 μL, Serum 50 μL, Supernatant 50 μL | |
NOTCH3 | |
4 hrs 45 mins |
0.05 to 10 ng/mL | |
HRP | |
Plasma, Serum, Supernatant | |
4854 | |
<12% | |
Pre-coated 96 well plate, Standard, Assay Diluent concentrate, Biotinylated Detection Antibody, SAV-HRP, Wash Buffer, Chromogen, Stop Solution, Adhesive Plate Covers | |
RUO | |
2°C to 8°C | |
1 hrs 20 mins |
Individuate un'opportunità di miglioramento?Condividi una correzione di contenuto
Correzione del contenuto del prodotto
Fornite il vostro feedback sul contenuto del prodotto compilando il modulo sottostante.
Titolo del prodotto