Learn More
Invitrogen™ Human IL-23 R ELISA Kit
ELISA
Specifica
Numero di accesso | IBD17; IL 23 R; IL-23 receptor; Il23r; IL-23 R; interleukin 23 receptor; interleukin 23 receptor-like; interleukin-23 receptor |
---|---|
Intervallo saggio | 0.2 to 50 ng/mL |
Assay Sensitivity | 0.2 ng/mL |
Coniugato | HRP |
Product Type | ELISA |
Descrizione
Human IL-23 R quantitates human IL-23 R in serum, plasma, supernatant. The assay will exclusively recognize both natural and recombinant human IL-23 R.
IL-23R is a 629 aa, novel subunit of the receptor for cytokine IL23A/IL23 which pairs with the receptor molecule IL12RB1/IL12R beta 1and thus take part in IL23A signaling. Signal transduction analysis confirms that this protein associates constitutively with Janus kinase 2 (JAK2), and also binds to transcription activator STAT3 in a ligand-dependent manner. Alternative splicing results in six spliced isoforms of IL-23R. Reports suggest that IL-23R plays an important role in psoriasis pathogenesis and is also a potential therapeutic target in inflammatory bowel disease.Specifica
IBD17; IL 23 R; IL-23 receptor; Il23r; IL-23 R; interleukin 23 receptor; interleukin 23 receptor-like; interleukin-23 receptor | |
0.2 ng/mL | |
ELISA | |
Colorimetric Microplate Reader | |
Il23r | |
<10 | |
HRP | |
Plasma 50 μL, Serum 50 μL, Supernatant 50 μL | |
IL23R | |
4 hrs 45 mins |
0.2 to 50 ng/mL | |
HRP | |
Plasma, Serum, Supernatant | |
149233 | |
<12% | |
Pre-coated 96 well plate, Standard, Assay Diluent concentrate, Biotinylated Detection Antibody, SAV-HRP, Wash Buffer, Chromogen, Stop Solution, Adhesive Plate Covers | |
RUO | |
2°C to 8°C | |
1 hrs 20 mins |