missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ Human FLRT3 ELISA Kit
ELISA
Specifica
Numero di accesso | 5530600M07Rik; C430047I10Rik; Fibronectin leucine rich transmembrane protein 3; Fibronectin-like domain-containing leucine-rich transmembrane protein 3; FLRT3; HH21; KIAA1469; Leucine-rich repeat transmembrane protein FLRT3; mKIAA1469; RGD1566005; UNQ856/PRO1865 |
---|---|
Intervallo saggio | 0.14 to 25 ng/mL |
Assay Sensitivity | 0.14 ng/mL |
Coniugato | HRP |
Product Type | ELISA |
Descrizione
Human FLRT3 quantitates human FLRT3 in serum, plasma, supernatant. The assay will exclusively recognize both natural and recombinant human FLRT3.
This gene encodes a member of the fibronectin leucine rich transmembrane protein (FLRT) family. FLRTs may function in cell adhesion and/or receptor signaling. Their protein structures resemble small leucine-rich proteoglycans found in the extracellular matrix. This gene is expressed in many tissues. Two alternatively spliced transcript variants encoding the same protein have been described for this gene.Specifica
5530600M07Rik; C430047I10Rik; Fibronectin leucine rich transmembrane protein 3; Fibronectin-like domain-containing leucine-rich transmembrane protein 3; FLRT3; HH21; KIAA1469; Leucine-rich repeat transmembrane protein FLRT3; mKIAA1469; RGD1566005; UNQ856/PRO1865 | |
0.14 ng/mL | |
ELISA | |
Colorimetric Microplate Reader | |
FLRT3 | |
<10 | |
HRP | |
Plasma 50 μL, Serum 50 μL, Supernatant 50 μL | |
FLRT3 | |
4 hrs 45 mins |
0.14 to 25 ng/mL | |
HRP | |
Plasma, Serum, Supernatant | |
23767 | |
<12% | |
Pre-coated 96 well plate, Standard, Assay Diluent concentrate, Biotinylated Detection Antibody, SAV-HRP, Wash Buffer, Chromogen, Stop Solution, Adhesive Plate Covers | |
RUO | |
2°C to 8°C | |
1 hrs 20 mins |
Individuate un'opportunità di miglioramento?Condividi una correzione di contenuto
Correzione del contenuto del prodotto
Fornite il vostro feedback sul contenuto del prodotto compilando il modulo sottostante.
Titolo del prodotto