missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ Human FABP-5 ELISA Kit
ELISA
712.00€ - 5420.00€
Specifications
Accession Number | C-FABP; Cutaneous fatty acid-binding protein; DA11; EFABP; E-FABP; E-FABPPAFABP; Epidermal-type fatty acid-binding protein; epithelial fatty acid-binding protein; FABP5; Fabpe; fatty acid binding protein 5; fatty acid binding protein 5 (psoriasis-associated); fatty acid binding protein 5, epidermal; fatty acid-binding protein 5; Fatty acid-binding protein, epidermal; fatty acid-binding protein, epidermal; uncharacterized protein LOC16592; Keratinocyte lipid-binding protein; KFABP; Klbp; Mal1; PAFABP; PA-FABP; PA-FABPepidermal; psoriasis-associated fatty acid-binding protein homolog |
---|---|
Assay Range | 12.5 to 800 ng/mL |
Assay Sensitivity | 12.5 ng/mL |
Conjugate | HRP |
Product Type | ELISA |
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
17164929
|
Invitrogen™
EH178RBX5 |
5 x 96 Tests |
2860.00€
Each |
Por favor, inicie sesión para comprar este producto. ¿No tiene usuario web? Regístrese hoy mismo. | |||||
17804090
|
Invitrogen™
EH178RBX10 |
10 x 96 Tests |
5420.00€
Pack of 10 |
Por favor, inicie sesión para comprar este producto. ¿No tiene usuario web? Regístrese hoy mismo. | |||||
17114463
|
Invitrogen™
EH178RB |
96 Tests |
712.00€
Pack of 96 |
Por favor, inicie sesión para comprar este producto. ¿No tiene usuario web? Regístrese hoy mismo. | |||||
Description
Human FABP-5 quantitates human FABP-5 in serum, plasma, supernatant. The assay will exclusively recognize both natural and recombinant human FABP-5.
FABP5 (fatty acid binding protein 5 (psoriasis-associated)) is a proteinthat in humans is encoded by theFABP5 gene, also known as PAFABP, EFABP, E-FABP, KFABP, PA-FABP, Epidermal-type fatty acid-binding protein, Fatty acid-binding protein 5, Psoriasis-associated fatty acid-binding protein homolog. It is a family of small, highly conserved, cytoplasmic proteins that bind long-chain fatty acids and other hydrophobic ligands. The PAFABP cDNA encodes a 135-amino acid protein with molecular weight 15,164. This gene encodes the fatty acid binding protein found in epidermal cells, and was first identified as being upregulated inpsoriasistissue. It is though that FABPs roles include fatty acid uptake, transport, and metabolism. Kaczocha et al. (2009) identifiedFABP5and FABP7 as cytosolic proteins that transport AEA from the plasma membrane to subcellular fatty acid amide hydrolase, where it is hydrolyzed and inactivated. FABP3 did not show this specific transport function.Specifications
C-FABP; Cutaneous fatty acid-binding protein; DA11; EFABP; E-FABP; E-FABPPAFABP; Epidermal-type fatty acid-binding protein; epithelial fatty acid-binding protein; FABP5; Fabpe; fatty acid binding protein 5; fatty acid binding protein 5 (psoriasis-associated); fatty acid binding protein 5, epidermal; fatty acid-binding protein 5; Fatty acid-binding protein, epidermal; fatty acid-binding protein, epidermal; uncharacterized protein LOC16592; Keratinocyte lipid-binding protein; KFABP; Klbp; Mal1; PAFABP; PA-FABP; PA-FABPepidermal; psoriasis-associated fatty acid-binding protein homolog | |
12.5 ng/mL | |
ELISA | |
Colorimetric Microplate Reader | |
Fabp5 | |
<10 | |
HRP | |
Plasma 50 μL, Serum 50 μL, Supernatant 50 μL | |
FABP5 | |
4 hrs 45 mins |
12.5 to 800 ng/mL | |
HRP | |
Plasma, Serum, Supernatant | |
2171 | |
<12% | |
Pre-coated 96 well plate, Standard, Assay Diluent concentrate, Biotinylated Detection Antibody, SAV-HRP, Wash Buffer, Chromogen, Stop Solution, Adhesive Plate Covers | |
RUO | |
2°C to 8°C | |
1 hrs 20 mins |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title