missing translation for 'onlineSavingsMsg'
Learn More

Invitrogen™ Human Dnmt3b ELISA Kit

ELISA

Specifica

Numero di accesso DNA (cytosine-5-)-methyltransferase 3 beta; DNA (cytosine-5)-methyltransferase 3 B; DNA methyltransferase 3 beta; DNA methyltransferase 3 B; DNA methyltransferase HsaIIIB; DNA methyltransferase MmuIIIB; DNA MTase HsaIIIB; DNA MTase MmuIIIB; Dnmt3; DNMT3B; ICF; ICF1; M.HsaIIIB; m.MmuIIIB; MGC1090; MmuIIIB
Intervallo saggio 0.64 to 150 ng/mL
Assay Sensitivity 0.64 ng/mL
Coniugato HRP
Product Type ELISA
Visualizza altre specifiche

Descrizione

Descrizione

Human Dnmt3b quantitates human Dnmt3b in serum, plasma, supernatant. The assay will exclusively recognize both natural and recombinant human Dnmt3b.

CpG methylation is an epigenetic modification that is important for embryonic development, imprinting, and X-chromosome inactivation. Studies in mice have demonstrated that DNA methylation is required for mammalian development. This gene encodes a DNA methyltransferase which is thought to function in de novo methylation, rather than maintenance methylation. The protein localizes primarily to the nucleus and its expression is developmentally regulated. Mutations in this gene cause the immunodeficiency-centromeric instability-facial anomalies (ICF) syndrome. Six alternatively spliced transcript variants have been described. The full length sequences of variants 4 and 5 have not been determined.
Specifica

Specifica

DNA (cytosine-5-)-methyltransferase 3 beta; DNA (cytosine-5)-methyltransferase 3 B; DNA methyltransferase 3 beta; DNA methyltransferase 3 B; DNA methyltransferase HsaIIIB; DNA methyltransferase MmuIIIB; DNA MTase HsaIIIB; DNA MTase MmuIIIB; Dnmt3; DNMT3B; ICF; ICF1; M.HsaIIIB; m.MmuIIIB; MGC1090; MmuIIIB
0.64 ng/mL
ELISA
Colorimetric Microplate Reader
Dnmt3b
<10
HRP
Plasma 50 μL, Serum 50 μL, Supernatant 50 μL
DNMT3B
4 hrs 45 mins
0.64 to 150 ng/mL
HRP
Plasma, Serum, Supernatant
1789
<12%
Pre-coated 96 well plate, Standard, Assay Diluent concentrate, Biotinylated Detection Antibody, SAV-HRP, Wash Buffer, Chromogen, Stop Solution, Adhesive Plate Covers
RUO
2°C to 8°C
1 hrs 20 mins
Videos
SDS
Documenti

Documenti

Product Certifications
Correzione del contenuto del prodotto

Fornite il vostro feedback sul contenuto del prodotto compilando il modulo sottostante.

Titolo del prodotto

Facendo clic su Invia, l'utente riconosce che potrebbe essere contattato da Fisher Scientific in merito al feedback fornito in questo modulo. Non condivideremo le vostre informazioni per altri scopi. Tutte le informazioni di contatto fornite saranno conservate in conformità con la nostra Politica sulla privacy. Informativa sulla privacy.

Grazie per averci aiutato a migliorare il nostro sito web. Il vostro feedback è stato inviato