missing translation for 'onlineSavingsMsg'
Learn More

Invitrogen™ Human DLEC/CLEC4C/BDCA-2 ELISA Kit

ELISA

Specifica

Numero di accesso BDCA2; BDCA-2; Blood dendritic cell antigen 2; blood dendritic cell antigen 2 protein; CD303; CD303 antigen; CLEC4C; CLECSF11; CLECSF7; C-type (calcium dependent, carbohydrate-recognition domain) lectin, superfamily member 11; C-type (calcium dependent, carbohydrate-recognition domain) lectin, superfamily member 7; C-type lectin domain family 4 member C; C-type lectin domain family 4, member C; C-type lectin domain family 4, member C, isoform 1; C-type lectin superfamily member 7; dendritic cell lectin b; dendritic lectin; DLEC; HECL; PRO34150; UNQ9361/PRO34150
Intervallo saggio 0.082 to 20 ng/mL
Assay Sensitivity 0.082 ng/mL
Coniugato HRP
Product Type ELISA
Visualizza altre specifiche

Descrizione

Descrizione

Human DLEC/CLEC4C/BDCA-2 quantitates human DLEC/CLEC4C/BDCA-2 in serum, plasma, supernatant. The assay will exclusively recognize both natural and recombinant human DLEC/CLEC4C/BDCA-2.

This gene encodes a member of the C-type lectin/C-type lectin-like domain superfamily. Members of this family share a common protein fold and have diverse functions, such as cell adhesion, cell-cell signalling, glycoprotein turnover, and roles in inflammation and immune response. The encoded type 2 transmembrane protein may play a role in dendritic cell function. Two transcript variants encoding distinct isoforms have been identified for this gene.
Specifica

Specifica

BDCA2; BDCA-2; Blood dendritic cell antigen 2; blood dendritic cell antigen 2 protein; CD303; CD303 antigen; CLEC4C; CLECSF11; CLECSF7; C-type (calcium dependent, carbohydrate-recognition domain) lectin, superfamily member 11; C-type (calcium dependent, carbohydrate-recognition domain) lectin, superfamily member 7; C-type lectin domain family 4 member C; C-type lectin domain family 4, member C; C-type lectin domain family 4, member C, isoform 1; C-type lectin superfamily member 7; dendritic cell lectin b; dendritic lectin; DLEC; HECL; PRO34150; UNQ9361/PRO34150
0.082 ng/mL
ELISA
Colorimetric Microplate Reader
CLEC4C
<10
HRP
Plasma 50 μL, Serum 50 μL, Supernatant 50 μL
CLEC4C
4 hrs 45 mins
0.082 to 20 ng/mL
HRP
Plasma, Serum, Supernatant
170482
<12%
Pre-coated 96 well plate, Standard, Assay Diluent concentrate, Biotinylated Detection Antibody, SAV-HRP, Wash Buffer, Chromogen, Stop Solution, Adhesive Plate Covers
RUO
2°C to 8°C
1 hrs 20 mins
Videos
SDS
Documenti

Documenti

Product Certifications
Correzione del contenuto del prodotto

Fornite il vostro feedback sul contenuto del prodotto compilando il modulo sottostante.

Titolo del prodotto

Facendo clic su Invia, l'utente riconosce che potrebbe essere contattato da Fisher Scientific in merito al feedback fornito in questo modulo. Non condivideremo le vostre informazioni per altri scopi. Tutte le informazioni di contatto fornite saranno conservate in conformità con la nostra Politica sulla privacy. Informativa sulla privacy.

Grazie per averci aiutato a migliorare il nostro sito web. Il vostro feedback è stato inviato