missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ Human Cyr61/CCN1 ELISA Kit
ELISA
Specifica
Numero di accesso | 3 CH61; AI325051; CCN family member 1; Ccn1; Cellular communication network factor 1; Cyr61; cysteine rich angiogenic inducer 61; cysteine rich protein 61; cysteine rich protein 61-like; cysteine-rich angiogenic inducer 61; cysteine-rich heparin-binding protein 61; cysteine-rich, angiogenic inducer, 61; cysteine-rich, anigogenic inducer, 61; GIG1; IBP-10; IGF-binding protein 10; IGFBP10; IGFBP-10; insulin-like growth factor binding protein 10; insulin-like growth factor-binding protein 10; intermediate early; LOC100339657; protein CYR61; Protein GIG1 |
---|---|
Intervallo saggio | 0.137 to 100 ng/mL |
Assay Sensitivity | 0.13 ng/mL |
Coniugato | HRP |
Product Type | ELISA |
Descrizione
Human Cyr61/CCN1 quantitates human Cyr61/CCN1 in serum, plasma, supernatant. The assay will exclusively recognize both natural and recombinant human Cyr61/CCN1.
CYR61 is a secreted heparin binding protein, encoded by a growth factor inducible immediate-early gene that associates with the extracellular matrix and connective tissue. CYR61 is a member of a distinct family of angiogenic and vasculogenic regulators designated CCN proteins, which includes connective tissue growth factor (CTGF) and the mouse CYR61 homolog, Fisp12. As an angiogenic inducer, CYR61 binds to the cell surface receptor integrin alphavbeta3, where it then stimulates cell adhesion and migration and promotes DNA synthesis of human vascular endothelial cells. Expression of CYR61 is elevated during vessel growth, wound healing and chondrocyte differentiation. CYR61 is also detected in a wide variety of tumors as it induces tumor growth and functions as a marker of tumor progression.Specifica
3 CH61; AI325051; CCN family member 1; Ccn1; Cellular communication network factor 1; Cyr61; cysteine rich angiogenic inducer 61; cysteine rich protein 61; cysteine rich protein 61-like; cysteine-rich angiogenic inducer 61; cysteine-rich heparin-binding protein 61; cysteine-rich, angiogenic inducer, 61; cysteine-rich, anigogenic inducer, 61; GIG1; IBP-10; IGF-binding protein 10; IGFBP10; IGFBP-10; insulin-like growth factor binding protein 10; insulin-like growth factor-binding protein 10; intermediate early; LOC100339657; protein CYR61; Protein GIG1 | |
0.13 ng/mL | |
ELISA | |
Colorimetric Microplate Reader | |
CCN1 | |
<10 | |
HRP | |
Plasma 50 μL, Serum 50 μL, Supernatant 50 μL | |
CYR61 | |
4 hrs 45 mins |
0.137 to 100 ng/mL | |
HRP | |
Plasma, Serum, Supernatant | |
3491 | |
<12% | |
Pre-coated 96 well plate, Standard, Assay Diluent concentrate, Biotinylated Detection Antibody, SAV-HRP, Wash Buffer, Chromogen, Stop Solution, Adhesive Plate Covers | |
RUO | |
2°C to 8°C | |
1 hrs 20 mins |
Individuate un'opportunità di miglioramento?Condividi una correzione di contenuto
Correzione del contenuto del prodotto
Fornite il vostro feedback sul contenuto del prodotto compilando il modulo sottostante.
Titolo del prodotto