missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ Human CLEC9A ELISA Kit
ELISA
Specifica
Numero di accesso | 9830005G06Rik; CD370; CLEC9A; C-type lectin domain family 9 member A; C-type lectin domain family 9, member A; dendritic cell natural killer lectin group receptor 1; DNGR1; Dngr-1; HEEE9341; RGD1562513; UNQ9341; UNQ9341/PRO34046 |
---|---|
Intervallo saggio | 0.12 to 25 ng/mL |
Assay Sensitivity | 0.12 ng/mL |
Coniugato | HRP |
Product Type | ELISA |
Descrizione
Human CLEC9A quantitates human CLEC9A in serum, plasma, supernatant. The assay will exclusively recognize both natural and recombinant human CLEC9A.
CLEC9A is a group V C-type lectin-like receptor (CTLR) that functions as an activation receptor and is expressed on myeloid lineage cells.Specifica
9830005G06Rik; CD370; CLEC9A; C-type lectin domain family 9 member A; C-type lectin domain family 9, member A; dendritic cell natural killer lectin group receptor 1; DNGR1; Dngr-1; HEEE9341; RGD1562513; UNQ9341; UNQ9341/PRO34046 | |
0.12 ng/mL | |
ELISA | |
Colorimetric Microplate Reader | |
CLEC9A | |
<10 | |
HRP | |
Plasma 50 μL, Serum 50 μL, Supernatant 50 μL | |
CLEC9A | |
4 hrs 45 mins |
0.12 to 25 ng/mL | |
HRP | |
Plasma, Serum, Supernatant | |
283420 | |
<12% | |
Pre-coated 96 well plate, Standard, Assay Diluent concentrate, Biotinylated Detection Antibody, SAV-HRP, Wash Buffer, Chromogen, Stop Solution, Adhesive Plate Covers | |
RUO | |
2°C to 8°C | |
1 hrs 20 mins |
Individuate un'opportunità di miglioramento?Condividi una correzione di contenuto
Correzione del contenuto del prodotto
Fornite il vostro feedback sul contenuto del prodotto compilando il modulo sottostante.
Titolo del prodotto