missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ Human CD177 ELISA Kit
ELISA
712.00€ - 2860.00€
Specifications
Accession Number | 1190003K14Rik; Cd177; CD177 antigen; CD177 molecule; cell surface receptor; HNA2A; HNA-2 A; Human neutrophil alloantigen 2 A; NB1; NB1 glycoprotein; NB1 GP; Pdp3; Polycythemia rubra vera protein 1; PRV1; PRV-1; RGD1562941; UNQ595/PRO1181 |
---|---|
Assay Range | 0.21 to 50 ng/mL |
Assay Sensitivity | 0.21 ng/mL |
Conjugate | HRP |
Product Type | ELISA |
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
17105319
|
Invitrogen™
EH80RBX5 |
5 x 96 Tests |
2860.00€
Each |
Por favor, inicie sesión para comprar este producto. ¿No tiene usuario web? Regístrese hoy mismo. | |||||
17144833
|
Invitrogen™
EH80RB |
96 Tests |
712.00€
Pack of 96 |
Por favor, inicie sesión para comprar este producto. ¿No tiene usuario web? Regístrese hoy mismo. | |||||
Description
Human CD177 quantitates human CD177 in serum, plasma, supernatant. The assay will exclusively recognize both natural and recombinant human CD177.
CD177 (NB1/HNA-2a and PRV-1 form) is a GPI-anchored glycoprotein present mainly on neutrophils. Its plasma membrane expression is increased during pregnancy and and inflammation or after G-CSF application. Ligand of CD177 has been identified as CD31 (PECAM-1). CD177 participates in neutrophil transmigration and seems to be also a pro-proliferative molecule. The antibodies against CD177 can be involved in neonatal alloimmune neutropenia (NAN).Specifications
1190003K14Rik; Cd177; CD177 antigen; CD177 molecule; cell surface receptor; HNA2A; HNA-2 A; Human neutrophil alloantigen 2 A; NB1; NB1 glycoprotein; NB1 GP; Pdp3; Polycythemia rubra vera protein 1; PRV1; PRV-1; RGD1562941; UNQ595/PRO1181 | |
0.21 ng/mL | |
ELISA | |
Colorimetric Microplate Reader | |
CD177 | |
<10 | |
HRP | |
Plasma 50 μL, Serum 50 μL, Supernatant 50 μL | |
CD177 | |
4 hrs 45 mins |
0.21 to 50 ng/mL | |
HRP | |
Plasma, Serum, Supernatant | |
57126 | |
<12% | |
Pre-coated 96 well plate, Standard, Assay Diluent concentrate, Biotinylated Detection Antibody, SAV-HRP, Wash Buffer, Chromogen, Stop Solution, Adhesive Plate Covers | |
RUO | |
2°C to 8°C | |
1 hrs 20 mins |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title