Learn More
Buerkle™ Graduated Polypropylene Cylinder with Embossed Gradations
Read molded-in graduations with ease to ensure accuracy with the Buerkle™ Graduated Polypropylene Cylinder with Embossed Gradations. This graduated cylinder offers excellent chemical resistance for a wide variety of solutions.
Marca: Buerkle™ 7502-0250
Questo articolo non è restituibile.
Consulta la politica di reso
Descrizione
- Resists temperatures up to 135°C (275°F)
- Autoclavable
- Individually packaged
- Polypropylene plastic
Specifica
250 mL | |
Category B DIN 12681/ISO 6706 | |
75 mm | |
6/Pk. | |
Sì | |
Chemists, During Outdoor Field Work and Wherever Volumetric Measurements Have to Be Made During Production, Laboratories, Workshop | |
Esagonale | |
Cilindro graduato |
PP (polipropilene) | |
Embossed | |
Embossed graduation | |
2 mL | |
Round | |
Blu | |
315 mm |