Learn More
Buerkle™ Graduated Polypropylene Cylinder with Embossed Gradations
Read molded-in graduations with ease to ensure accuracy with the Buerkle™ Graduated Polypropylene Cylinder with Embossed Gradations. This graduated cylinder offers excellent chemical resistance for a wide variety of solutions.
Marca: Buerkle™ 7502-0050
Questo articolo non è restituibile.
Consulta la politica di reso
Descrizione
- Resists temperatures up to 135°C (275°F)
- Autoclavable
- Individually packaged
- Polypropylene plastic
Specifica
50 ml | |
Category B DIN 12681/ISO 6706 | |
27 mm | |
1 | |
Sì | |
Chemists, During Outdoor Field Work and Wherever Volumetric Measurements Have to Be Made During Production, Laboratories, Workshop | |
Esagonale | |
Cilindro graduato |
PP (polipropilene) | |
Embossed | |
Embossed graduation | |
1 mL | |
Round | |
Blu | |
200 mm |
Titolo del prodotto
Buerkle™ Graduated Polypropylene Cylinder with Embossed Gradations > Capacity: 50mL; Division: 1mL; Height: 200mm