Learn More
Fisherbrand™ Flask Clamp for Shaking Incubators
Fisherbrand™ Flask Clamp is designed to be used with a shaking incubator and can hold a variety of flask sizes.
Brand:  Fisherbrand™ H1000-MR-250
49.81 EUR valid until 2025-12-31
Use promo code "25339" to get your promotional price.
 View more versions of this product
Product Code. 16313236
  • 80.85€ / Each
Only null left
Estimated Shipment: 19-08-2025
Add to basket
Description
Description
  • Constructed of stainless steel
  • For use with shaking incubators
  • Designed to hold flasks
Specifications
Specifications
250 mL Flasks
Stainless Steel
1 Ea.
Flask Clamp
16313226
Product Suggestions
Product Suggestions
Videos
SDS
Documents
Documents
Certificates
Promotions
Promotions