Learn More
Fisherbrand™ Vortex Mixer Head
For use with Fisherbrand vortex mixers.
Marca:  Fisherbrand™ 80862999
27.81 EUR valido fino al 2025-12-31
Usa il codice promo "25339" per avere il tuo prezzo promozionale.
Codice prodotto. 15756949
  • 36.15€ / Pezzo
Solo null disponibili
Spedizione stimata: 03-09-2025
Aggiungi al carrello
Descrizione
Descrizione
Also compatible with other manufacturers' models; contact your Fisher Scientific Customer Service Representative for details.
  • Rubber head (76mm) 02-215-382 is used with 76mm head cover 02-215-383 for mixing beakers, flasks, and irregular shaped objects
Also compatible with other manufacturers' models; contact your Fisher Scientific Customer Service Representative for details.
  • Rubber head (76mm) 02-215-382 is used with 76mm head cover 02-215-383 for mixing beakers, flasks, and irregular shaped objects
TRUSTED_SUSTAINABILITY
Specifica
Specifica
Head
76 mm
Vortex mixers