Learn More
Deltalab™ Eurotubo Polypropylene Centrifuge Tubes with Caps
Eurotubo Polypropylene Centrifuge Tubes with Green Caps are available sterile or non-sterile. The external skirt allows the tubes to remain free-standing. Caps should be loosened when autoclaving.
Brand: Deltalab™ 401402
This item is not returnable.
View return policy
Description
- Samples and/or urine collection containers
- Autoclavable
- HDPE green cap
- External skirt
- Pre-fitted cap
Specifications
Green | |
12 mL | |
1500/Cs. | |
Case | |
Autoclavable | |
Screw Cap | |
Centrifuge Tube |
Clear | |
Polypropylene | |
250 Pk. | |
Sterile | |
102.9 mm | |
16.2 mm | |
15,000 x g |