Learn More
EEF1D Mouse anti-Human, CoraLite 594, Clone: 3B1B11, Proteintech
Mouse Monoclonal Antibody
Marca:  Proteintech CL594-60085-100UL
Codice prodotto. 16890876
  • 450.00€ / 100µL
Questo articolo non è restituibile. Consulta la politica di reso
Descrizione
Descrizione
The EEF1D antibody from Proteintech is a mouse monoclonal antibody generated with recombinant protein of human EEF1D. This antibody recognizes human, mouse, rat antigen. The EEF1D antibody has been validated for the following applications: IF analysis. This antibody is coralite™594.
Specifica
Specifica
EEF1D
Monoclonal
2000 μg/mL
Mouse
Primary
Human, Rat, Mouse
Immunofluorescence
3B1B11
CoraLite 594
100 μL
1936
IgG1
Product Suggestions
Product Suggestions
Videos
SDS
Documents
Documents
Certificates
Promotions
Promotions
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.
Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.