Learn More
Lenz Laborglasinstrumente™ Tubo filettato in vetro Duran™
Provides for secure and easy leak-tight connection of apparatus in the laboratory. Lenz Laborglasinstrumente™ Screwthread Tubes with NS Cone are manufactured from durable DURAN™ borosilicate glass.
Marca: Lenz Laborglasinstrumente™ 01311829
Questo articolo non è restituibile.
Consulta la politica di reso
Descrizione
- Acc. to DIN 12257
- Available in a range of socket and thread sizes
- Minimized need for grease
- Corrosion-free and resistant to thermal shock
- With conical joint, featuring high taper accuracy and excellent tightness
Specifica
Confezione da 1 | |
NS 29/32 | |
GL 18 | |
Duran Glass |