Learn More
Rogo Sampaic™ Double Clamps
Provides for secure rod/stem construction. Rogo Sampaic™ Double Clamps are manufactured from a durable nickel and chromium alloy.
Marca: Rogo Sampaic™ FCP019
Questo articolo non è restituibile.
Consulta la politica di reso
Descrizione
- Fits rods of 16mm diameters
- Sold individually
Specifica
Each | |
Chromium, Nickel | |
16 mm |
Double Clamp | |
perpendicular structure |