Learn More
Borer Chemie™ deconex™ 16 NT Liquid Cleaning Concentrate
For pre-cleaning and precision cleaning of medical instruments. Borer Chemie™ deconex™ 16 NT Liquid Cleaning Concentrate removes organic and inorganic contaminants from sensitive, metallic substrates.
Brand:  Borer Chemie™ D16NT.1000
Product Code. 10075160
This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products »
Explore more special offers
Description
Description
  • Emulsification activity spectrum
  • Ultrasonic
  • Compatible with aluminum, zinc, HSS/steel, sintered metals, stainless steel/Inox, nickel/titanium, silver, gold, platinum and hard metals
Specifications
Specifications
7.9
1.06 g/mL
1 kg
Product Suggestions
Product Suggestions
Videos
Safety and Handling
Safety and Handling
Product Identifier
  • deconex 16 NT
Signal Word
  • Danger
Hazard Category
  • Serious eye damage/eye irritation Category 1
  • Skin corrosion/irritation Category 2
Hazard Statement
  • H318-Causes serious eye damage.
  • H315-Causes skin irritation.
Precautionary Statement
  • P264-Wash hands thoroughly after handling.
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
  • P310-Immediately call a POISON CENTER/doctor/.
  • P302+P352-IF ON SKIN: Wash with plenty of water/and soap.
SDS
Documents
Documents
Certificates
Special Offers
Special Offers