missing translation for 'onlineSavingsMsg'
Learn More

Tocris Bioscience™ DAPT
GREENER_CHOICE

Product Code. 18024422
Click to view available options
Quantità:
10 mg
50 mg
Unit Size:
10mg
50mg
This item is not returnable. View return policy
This item is not returnable. View return policy

Gamma γ-secretase inhibitor; induces neuronal differentiation; blocks Notch signaling

DAPT is a γ-secretase inhibitor. DAPT reduces Aβ40 and Aβ42 levels in human primary neuronal cultures (IC50 values are 115 and 200 nM for total Aβ and Aβ42 respectively) and in brain extract, cerebrospinal fluid and plasma in vivo. DAPT has no effect on APPα and APPβ levels. DAPT blocks Notch signaling in hybrid human-mouse fetal thymus organ culture (FTOC) and causes ESCs to commit to neuronal differentiation. DAPT can be used in a small molecule cocktail to derive cortical neurons from hPSCs and to maintain hepatocytes in culture. DAPT also promotes the formation of cone photoreceptors in retinal organoids.

For more information about how DAPT may be used, see our protocols: Accelerated Induction of Cortical Neurons from hiPSCs, Human iPSC-derived Myoblasts, Generating Midbrain Dopaminergic Neurons from hPSCs .

DAPT synthesized to Ancillary Material Grade also available.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 208255-80-5
Formula molecolare C23H26F2N2O4
Peso molecolare 432.468

Specifications

Materiale o nome chimico DAPT
Quantità 50 mg
Peso formulazione 432.46
Purezza >99%
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.