Learn More
Thermo Scientific™ Culti-Loops™ Escherichia coli ATCC™ 51755™
Ready-to-use QC organisms recommended for use in performance testing of media, stains, reagents and identification kits, and for the evaluation of bacteriological procedures.
Marca:  Thermo Scientific™ R4601998
Codice prodotto. 12938968
Questo articolo non è attualmente disponibile o è stato dismesso.
La preghiamo di consultare la pagina del prodotto per individuare le possibili alternative.
Visualizzare prodotti alternativi »
Questo articolo non è restituibile. Consulta la politica di reso
Descrizione
Descrizione
Thermo Scientific™ Culti-Loops™ are ready-to-use QC organisms recommended for use in performance testing of media, stains, reagents and identification kits, and for the evaluation of bacteriological procedures.
The loop may be dissolved in liquid medium, Culti-Loops Aerobic Set Rehydration Fluid, Part No. R4631000 or streaked directly onto an appropriate media.
Ordering Information:
Culti-Loops are individually sealed in a foil pouch and packaged five loops per package.
TRUSTED_SUSTAINABILITY
Specifica
Specifica
51755TM*
Culti-Loops
Gram-negative Rods
5 Loops/Pack
Escherichia coli ATCC™ 51755™
Culti-Loops Quality Control Organism
Qualitative QC
BDMS 605
Certificati
Promozioni
Promozioni
*The ATCC Licensed Derivative Emblem, the ATCC Licensed Derivative word mark, and the ATCC catalog marks are trademarks of ATCC™ and are used under license.