Learn More
Thermo Scientific™ Culti-Loops™ Candida albicans ATCC™ 90028™
Descrizione
Thermo Scientific™ Culti-Loops™ are ready-to-use QC organisms recommended for use in performance testing of media, stains, reagents and identification kits, and for the evaluation of bacteriological procedures.
The loop may be dissolved in liquid medium, Culti-Loops Aerobic Set Rehydration Fluid, Part No. R4631000 or streaked directly onto an appropriate media.
Ordering Information:
Culti-Loops are individually sealed in a foil pouch and packaged five loops per package.
Specifica
Specifica
ATCC® | 90028TM* |
Certificazione/Conformità | Culti-Loops are CE Marked. |
Descrizione | Candida albicans |
Product Type | Single Organism |
Target Organism Class | Fungi |
Test Type | Qualitative QC |
Tipo di imballaggio | 5 Loops/Pack |
Quantità | 5 Loops/Pk. |
Strain Designations | NCCLS 11 |
*The ATCC Licensed Derivative Emblem, the ATCC Licensed Derivative word mark, and the ATCC catalog marks are trademarks of ATCC™ and are used under license.