Learn More
Thermo Scientific™ Culti-Loops™ Aeromonas hydrophila ATCC™ 7966™
Ready-to-use QC organisms recommended for use in performance testing of media, stains, reagents and identification kits, and for the evaluation of bacteriological procedures.
Marca: Thermo Scientific™ R4601020
202.95 EUR valido fino al 2025-12-31
Usa il codice promo "25339" per avere il tuo prezzo promozionale.
Descrizione
Thermo Scientific™ Culti-Loops™ are ready-to-use QC organisms recommended for use in performance testing of media, stains, reagents and identification kits, and for the evaluation of bacteriological procedures.
The loop may be dissolved in liquid medium, Culti-Loops Aerobic Set Rehydration Fluid, Part No. R4631000 or streaked directly onto an appropriate media.
Ordering Information:
Culti-Loops are individually sealed in a foil pouch and packaged five loops per package.
Specifica
Aeromonas species | |
Aeromonas hydrophila | |
Culti-Loops Quality Control Organism | |
Qualitative QC | |
WDCM00036, [CDC 359-60, IAM 12460, NCIB 9240, NCMB 86, NCTC 8049, RH 250] |
7966TM* | |
Culti-Loops | |
Gram-negative Rods | |
5 Loops/Pack |
*The ATCC Licensed Derivative Emblem, the ATCC Licensed Derivative word mark, and the ATCC catalog marks are trademarks of ATCC™ and are used under license.