Learn More
Sartorius Cubis™ II Precision High-Capacity Balance, MCE User interface
BIOPHARMAGREENER_CHOICE
Cubis™ II Precision High-Capacity Balance features fully customizable hardware, software, and connectivity
Marca:  Sartorius MCE36201S0-S00
8759.00 EUR valido fino al 2025-12-31
Usa il codice promo "24377" per avere il tuo prezzo promozionale.
Codice prodotto. 16283272
  • 9220.00€ / 1 unità
Questo articolo non è restituibile. Consulta la politica di reso
Descrizione
Descrizione
  • Monolithical weighing system
  • Integrated climate sensors
  • Individual sample holders
  • Error-free operation
  • Individual QApp workflows
  • Motorized auto-leveling
  • Full end-to-end data integrity
  • Integrated audit-trail
  • State-of-the-art user management
  • Excellent service support
  • Integrated status center
  • Integrated service functions
  • Preventive maintenance on accredited standards
  • TFT touch screen operation with readable display and easy to operate user interface
  • Software Factory installed basic set of essential weighing applications
  • Operation Activated by touch key, touch-free using IR sensor (draft shield M), or gesture sensor (optional) learning capability
Servizi
10-year warranty
TRUSTED_SUSTAINABILITY
Specifica
Specifica
2 USB A ports, 1 USB B port, 1USP C port, 1-9 pin RS 232 connection, 1 ethernet port
TFT Touchscreen
Balance, Accessories, Power supply, Manual, Factory test report, QP99 software
No draft shield
1 sec
10 anni
400 mm
15.8/17.1 kg
100/240 V ±10%
Class III
36.2 kg
1 g
Die-cast Aluminum, Plastic PBT, Optiwhite Float Glass and Stainless Steel, Aluminum Trim
±20 mg
High Capacity Precision Balance
412 mm
126/159 mm
120/240 V
50/60 Hz