Learn More
Sartorius Cubis™ II Precision (1-place) Balance, MCE Interface
Cubis™ II Precision (1-Place) Balance features fully customizable hardware, software, and connectivity
Marca: Sartorius MCE12201S0-S00
5715.00 EUR valido fino al 2025-12-31
Usa il codice promo "25339" per avere il tuo prezzo promozionale.
Questo articolo non è restituibile.
Consulta la politica di reso
Descrizione
- Monolithical weighing system
- Integrated climate sensors
- Individual sample holders
- Error-free operation
- Individual QApp workflows
- Motorized auto-leveling
- Full end-to-end data integrity
- Integrated audit-trail
- State-of-the-art user management
- Excellent service support
- Integrated status center
- Integrated service functions
- Preventive maintenance on accredited standards
- TFT touch screen operation with readable display and easy to operate user interface
- Software Factory installed basic set of essential weighing applications
- Operation Activated by touch key, touch-free using IR sensor (draft shield M), or gesture sensor (optional) learning capability
Servizi
10-year warranty
Specifica
2 USB A ports, 1 USB B port, 1USP C port, 1-9 pin RS 232 connection, 1 ethernet port | |
TFT Touchscreen | |
Balance, Accessories, Power supply, Manual, Factory test report, QP99 Software | |
Die-cast Aluminum, Plastic PBT, Optiwhite Float Glass and Stainless Steel, Aluminum Trim | |
±0.05 g | |
Precision Balance | |
425 mm | |
95 mm | |
120/240 V | |
50/60 Hz (± 5%) |
12.2 kg | |
20 mg | |
±30 mg | |
No draft shield | |
0.8 s | |
10 anni | |
240 mm | |
5.4 kg | |
100/240 V | |
Class I |